The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280789.1 | NP_001267718.1 | 1 - 2 | 247 | 91 |
NM_001280792.1 | NP_001267721.1 | 1 - 2 | 247 | 91 | |
NM_001280793.1 | NP_001267722.1 | 1 - 2 | 247 | 91 | |
NM_033161.3 | NP_149351.1 | 1 - 2 | 247 | 91 | |
GenBank mRNA | AF078866.1 | - | 1 - 2 | 163 | 91 |
AF078867.1 | - | 1 - 2 | 163 | 91 | |
AK299656.1 | - | 1 - 2 | 141 | 91 | |
AK300004.1 | - | 1 - 2 | 141 | 91 | |
AK315488.1 | - | 1 - 2 | 201 | 91 | |
BC018741.1 | - | 1 - 2 | 70 | 91 | |
BC111023.1 | - | 1 - 2 | 112 | 91 | |
BM148022.1 | - | 1 - 2 | 181 | 91 | |
BX647961.1 | - | 1 - 2 | 100 | 91 | |
DA842525.1 | - | 1 - 2 | 134 | 91 | |
DC424374.1 | - | 1 - 2 | 249 | 91 | |
Y14820.1 | - | 1 - 2 | 29 | 91 |