The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033549.4 | NP_291027.3 | 1 - 2 | 1547 | 78 |
NM_201627.2 | NP_963921.1 | 1 - 2 | 1547 | 78 | |
NR_045218.1 | 1 - 2 | 1547 | 78 | ||
XM_006714929.2 | XP_006714992.1 | 1 - 2 | 1545 | 78 | |
XM_006714930.2 | XP_006714993.1 | 1 - 2 | 1545 | 78 | |
XM_006714931.3 | XP_006714994.1 | 1 - 2 | 1545 | 78 | |
XM_006714932.3 | XP_006714995.1 | 1 - 2 | 1545 | 78 | |
GenBank mRNA | AB063180.1 | - | 1 - 2 | 573 | 78 |
AB100366.1 | - | 1 - 2 | 810 | 78 | |
AB100367.1 | - | 1 - 2 | 810 | 78 | |
AK027601.1 | - | 1 - 2 | 1520 | 78 | |
AK074052.1 | - | 1 - 2 | 644 | 78 | |
AL832960.1 | - | 1 - 2 | 424 | 78 | |
AX775789.1 | - | 1 - 2 | 286 | 78 | |
AX775791.1 | - | 1 - 2 | 286 | 78 | |
AX775793.1 | - | 1 - 2 | 1520 | 78 | |
AX775797.1 | - | 1 - 2 | 578 | 78 | |
AX775799.1 | - | 1 - 2 | 1520 | 78 | |
AX775801.1 | - | 1 - 2 | 1520 | 78 | |
AX775803.1 | - | 1 - 2 | 578 | 78 | |
BC004956.1 | - | 1 - 2 | 752 | 78 | |
BC018765.1 | - | 1 - 2 | 578 | 78 | |
BC071887.1 | - | 1 - 2 | 752 | 78 | |
BC090953.1 | - | 1 - 2 | 1090 | 78 |