The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058246.3 | NP_490647.1 | 8 - 9 | 846 | 101 |
XM_005249515.2 | XP_005249572.1 | 8 - 9 | 838 | 101 | |
XM_005249516.2 | XP_005249573.1 | 8 - 9 | 982 | 101 | |
XM_011515704.1 | XP_011514006.1 | 8 - 9 | 817 | 101 | |
GenBank mRNA | AB015798.1 | - | 8 - 9 | 839 | 101 |
AB209859.1 | - | 8 - 9 | 840 | 101 | |
AK223601.1 | - | 8 - 9 | 857 | 101 | |
AK293025.1 | - | 8 - 9 | 830 | 101 | |
AK297796.1 | - | 7 - 8 | 737 | 101 | |
AK297989.1 | - | 8 - 9 | 706 | 101 | |
AK301152.1 | - | 6 - 7 | 685 | 101 | |
BC002446.2 | - | 8 - 9 | 846 | 101 |