The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080430.2 | NP_536355.1 | 1 - 2 | 193 | 62 |
GenBank mRNA | AY043487.1 | - | 1 - 2 | 222 | 62 |
BC013421.1 | - | 1 - 2 | 139 | 62 | |
BC030236.1 | - | 1 - 2 | 150 | 62 | |
BC068004.1 | - | 1 - 2 | 183 | 62 |