The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080657.4 | NP_542388.2 | 1 - 2 | 481 | 78 |
XM_011510414.2 | XP_011508716.1 | 2 - 3 | 1570 | 78 | |
GenBank mRNA | AF442151.1 | - | 1 - 2 | 369 | 78 |
AK312670.1 | - | 1 - 2 | 380 | 78 | |
BC017969.1 | - | 1 - 2 | 369 | 78 |