Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRD
Entrez Gene ID
5789
Gene Name
protein tyrosine phosphatase, receptor type D
Gene Aliases
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location
Chr.9: 8314246 - 10612840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040712.2 NP_001035802.1 9 - 10 1638 82
NM_001171025.1 NP_001164496.1 8 - 9 1617 82
NM_002839.3 NP_002830.1 21 - 22 2287 82
NM_130391.3 NP_569075.2 10 - 11 1647 82
NM_130392.3 NP_569076.2 10 - 11 1647 82
NM_130393.3 NP_569077.2 9 - 10 1629 82
XM_006716817.3 XP_006716880.1 20 - 21 2585 82
XM_006716823.2 XP_006716886.1 20 - 21 2579 82
XM_006716825.3 XP_006716888.1 20 - 21 2581 82
XM_006716827.3 XP_006716890.1 17 - 18 2539 82
XM_006716832.3 XP_006716895.1 18 - 19 2543 82
XM_006716833.3 XP_006716896.1 20 - 21 2575 82
XM_006716834.3 XP_006716897.1 17 - 18 2537 82
XM_006716835.3 XP_006716898.1 19 - 20 2566 82
XM_006716837.3 XP_006716900.1 19 - 20 2555 82
XM_006716838.3 XP_006716901.1 18 - 19 2548 82
XM_006716839.3 XP_006716902.1 17 - 18 2534 82
XM_011517992.2 XP_011516294.1 20 - 21 2578 82
XM_017014958.1 XP_016870447.1 20 - 21 2584 82
XM_017014959.1 XP_016870448.1 21 - 22 2656 82
XM_017014960.1 XP_016870449.1 19 - 20 2545 82
XM_017014961.1 XP_016870450.1 19 - 20 2529 82
XM_017014962.1 XP_016870451.1 11 - 12 1717 82
XM_017014963.1 XP_016870452.1 20 - 21 2584 82
XM_017014964.1 XP_016870453.1 20 - 21 2583 82
XM_017014965.1 XP_016870454.1 20 - 21 2586 82
XM_017014966.1 XP_016870455.1 19 - 20 2575 82
XM_017014967.1 XP_016870456.1 19 - 20 2570 82
XM_017014968.1 XP_016870457.1 20 - 21 2581 82
XM_017014969.1 XP_016870458.1 19 - 20 2566 82
XM_017014970.1 XP_016870459.1 20 - 21 2584 82
XM_017014971.1 XP_016870460.1 18 - 19 2555 82
XM_017014972.1 XP_016870461.1 20 - 21 2584 82
XM_017014973.1 XP_016870462.1 18 - 19 2551 82
XM_017014974.1 XP_016870463.1 20 - 21 2584 82
XM_017014975.1 XP_016870464.1 17 - 18 2540 82
XM_017014976.1 XP_016870465.1 20 - 21 2581 82
XM_017014977.1 XP_016870466.1 20 - 21 2582 82
XM_017014978.1 XP_016870467.1 20 - 21 2581 82
XM_017014979.1 XP_016870468.1 20 - 21 2583 82
XM_017014980.1 XP_016870469.1 20 - 21 2578 82
XM_017014981.1 XP_016870470.1 17 - 18 2534 82
XM_017014982.1 XP_016870471.1 20 - 21 2580 82
XM_017014983.1 XP_016870472.1 20 - 21 2580 82
XM_017014984.1 XP_016870473.1 19 - 20 2571 82
XM_017014985.1 XP_016870474.1 20 - 21 2577 82
XM_017014986.1 XP_016870475.1 20 - 21 2576 82
XM_017014987.1 XP_016870476.1 19 - 20 2558 82
XM_017014988.1 XP_016870477.1 19 - 20 2558 82
XM_017014989.1 XP_016870478.1 20 - 21 2576 82
XM_017014990.1 XP_016870479.1 20 - 21 2575 82
XM_017014991.1 XP_016870480.1 20 - 21 2575 82
XM_017014992.1 XP_016870481.1 19 - 20 2566 82
XM_017014993.1 XP_016870482.1 20 - 21 2573 82
XM_017014994.1 XP_016870483.1 20 - 21 2573 82
XM_017014995.1 XP_016870484.1 19 - 20 2570 82
GenBank mRNA AB208869.1 - 8 - 9 1464 82
AB211400.1 - 20 - 21 2255 82
BC106713.1 - 8 - 9 1565 82
BC106714.1 - 10 - 11 1595 82
BC106715.2 - 10 - 11 1595 82
BC106716.1 - 9 - 10 1577 82
L38929.1 - 11 - 12 1697 82
X54133.1 - 1 - 2 377 82

View Full Product Details