The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301821.1 | NP_001288750.1 | 6 - 7 | 748 | 100 |
NM_001301822.1 | NP_001288751.1 | 5 - 6 | 636 | 100 | |
NM_130768.2 | NP_570124.1 | 6 - 7 | 748 | 100 | |
GenBank mRNA | AF461259.1 | - | 6 - 7 | 714 | 100 |
AK093445.1 | - | 6 - 7 | 701 | 100 | |
BC028695.1 | - | 6 - 7 | 718 | 100 | |
BC034963.1 | - | 6 - 7 | 723 | 100 | |
BC068488.1 | - | 5 - 6 | 609 | 100 | |
BC126186.1 | - | 6 - 7 | 707 | 100 | |
BC126188.1 | - | 6 - 7 | 707 | 100 | |
BC144212.1 | - | 6 - 7 | 707 | 100 |