The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133259.3 | NP_573566.2 | 24 - 25 | 2683 | 68 |
XM_006711915.2 | XP_006711978.1 | 24 - 25 | 2692 | 68 | |
XM_006711916.3 | XP_006711979.1 | 24 - 25 | 2667 | 68 | |
XM_011532473.1 | XP_011530775.1 | 24 - 25 | 2667 | 68 | |
XM_017003117.1 | XP_016858606.1 | 24 - 25 | 2665 | 68 | |
GenBank mRNA | AK125781.1 | - | 24 - 25 | 2654 | 68 |
AK290016.1 | - | 24 - 25 | 2654 | 68 | |
AK299869.1 | - | 23 - 24 | 2455 | 68 | |
AY289212.1 | - | 24 - 25 | 2683 | 68 | |
BC010282.1 | - | 4 - 5 | 550 | 68 | |
BC050311.1 | - | 24 - 25 | 2634 | 68 | |
BC130285.1 | - | 24 - 25 | 2679 | 68 | |
M92439.1 | - | 23 - 24 | 2307 | 68 |