The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143688.2 | NP_001137160.1 | 6 - 7 | 834 | 77 |
NM_001323936.1 | NP_001310865.1 | 6 - 7 | 753 | 77 | |
NM_001323937.1 | NP_001310866.1 | 5 - 6 | 655 | 77 | |
NM_001323938.1 | NP_001310867.1 | 6 - 7 | 761 | 77 | |
NM_001323939.1 | NP_001310868.1 | 7 - 8 | 958 | 77 | |
NM_001323940.1 | NP_001310869.1 | 6 - 7 | 834 | 77 | |
NM_001323941.1 | NP_001310870.1 | 6 - 7 | 1229 | 77 | |
NM_001323943.1 | NP_001310872.1 | 6 - 7 | 1028 | 77 | |
NM_001323944.1 | NP_001310873.1 | 6 - 7 | 1028 | 77 | |
NM_001323945.1 | NP_001310874.1 | 6 - 7 | 1302 | 77 | |
NM_001323946.1 | NP_001310875.1 | 6 - 7 | 955 | 77 | |
NM_001323948.1 | NP_001310877.1 | 5 - 6 | 899 | 77 | |
NM_133375.4 | NP_588616.1 | 6 - 7 | 784 | 77 | |
XM_005254146.3 | XP_005254203.1 | 6 - 7 | 902 | 77 | |
GenBank mRNA | AB075835.1 | - | 5 - 6 | 491 | 77 |
AK095407.1 | - | 6 - 7 | 813 | 77 | |
AK295392.1 | - | 6 - 7 | 661 | 77 | |
BC022089.2 | - | 6 - 7 | 719 | 77 |