The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178075.1 | NP_001171546.1 | 10 - 11 | 1413 | 126 |
NM_001178076.1 | NP_001171547.1 | 8 - 9 | 1179 | 126 | |
NM_001178077.1 | NP_001171548.1 | 8 - 9 | 1112 | 126 | |
NM_001673.4 | NP_001664.3 | 10 - 11 | 1487 | 126 | |
NM_133436.3 | NP_597680.2 | 10 - 11 | 1742 | 126 | |
NM_183356.3 | NP_899199.2 | 11 - 12 | 1765 | 126 | |
GenBank mRNA | AK000379.1 | - | 10 - 11 | 1710 | 126 |
AK302189.1 | - | 10 - 11 | 1307 | 126 | |
AK302242.1 | - | 8 - 9 | 1060 | 126 | |
AK316224.1 | - | 8 - 9 | 1114 | 126 | |
BC008723.2 | - | 11 - 12 | 1592 | 126 | |
BC014621.2 | - | 10 - 11 | 1363 | 126 | |
BT007113.1 | - | 8 - 9 | 1236 | 126 | |
M15798.1 | - | 9 - 10 | 1283 | 126 |