The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142466.2 | NP_001135938.1 | 5 - 6 | 666 | 56 |
NM_133443.3 | NP_597700.1 | 5 - 6 | 690 | 56 | |
XM_017023790.1 | XP_016879279.1 | 2 - 3 | 163 | 56 | |
GenBank mRNA | AK094971.1 | - | 5 - 6 | 590 | 56 |
AY029173.2 | - | 5 - 6 | 674 | 56 | |
BC062555.1 | - | 5 - 6 | 639 | 56 |