Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Chromosome Location
Chr.9: 127451486 - 127503501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005373.3 NP_001005373.1 19 - 20 1794 66
NM_001005374.3 NP_001005374.1 18 - 19 1499 66
NM_138361.5 NP_612370.3 18 - 19 2053 66
XM_006717316.3 XP_006717379.1 19 - 20 1794 66
XM_017015283.1 XP_016870772.1 19 - 20 2793 66
XM_017015284.1 XP_016870773.1 19 - 20 1789 66
GenBank mRNA AK056305.1 - 7 - 8 644 66
AK091589.1 - 18 - 19 1801 66
AK226106.1 - 19 - 20 1780 66
AY358830.1 - 19 - 20 1759 66
BC009239.2 - 18 - 19 1499 66

View Full Product Details