The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138410.3 | NP_612419.1 | 4 - 5 | 751 | 100 |
NM_181472.2 | NP_852137.1 | 3 - 4 | 652 | 100 | |
GenBank mRNA | AF479263.2 | - | 4 - 5 | 751 | 100 |
AK055554.1 | - | 1 - 2 | 1472 | 100 | |
AK308878.1 | - | 3 - 4 | 350 | 100 | |
BC010116.2 | - | 4 - 5 | 533 | 100 | |
CD366110.1 | - | 1 - 2 | 100 |