Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN15
Entrez Gene ID
24146
Gene Name
claudin 15
Gene Aliases
-
Chromosome Location
Chr.7: 101232092 - 101238820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC010160.1 - 2 - 3 527 76

View Full Product Details