The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138452.2 | NP_612461.1 | 1 - 2 | 248 | 64 |
GenBank mRNA | AK058159.1 | - | 1 - 2 | 248 | 64 |