The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281301.1 | NP_001268230.1 | 20 - 21 | 2697 | 107 |
NM_004856.6 | NP_004847.2 | 21 - 22 | 2774 | 107 | |
NM_138555.3 | NP_612565.1 | 22 - 23 | 3086 | 107 | |
XM_005254796.2 | XP_005254853.1 | 23 - 24 | 3077 | 107 | |
XM_005254797.2 | XP_005254854.1 | 22 - 23 | 2995 | 107 | |
XM_005254798.2 | XP_005254855.1 | 22 - 23 | 2825 | 107 | |
XM_005254799.2 | XP_005254856.1 | 22 - 23 | 2765 | 107 | |
XM_011522238.1 | XP_011520540.1 | 22 - 23 | 2912 | 107 | |
XM_011522239.2 | XP_011520541.1 | 22 - 23 | 2937 | 107 | |
XM_011522240.1 | XP_011520542.1 | 14 - 15 | 1931 | 107 | |
XM_017022733.1 | XP_016878222.1 | 20 - 21 | 2411 | 107 | |
GenBank mRNA | AI131325.1 | - | 1 - 2 | 107 | |
AK303874.1 | - | 17 - 18 | 2168 | 107 | |
AK308322.1 | - | 20 - 21 | 2619 | 107 | |
BC017705.2 | - | 21 - 22 | 2854 | 107 | |
BC051826.1 | - | 21 - 22 | 2716 | 107 | |
BI093060.1 | - | 3 - 4 | 176 | 107 | |
X67155.2 | - | 20 - 21 | 2614 | 107 |