The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014001.4 | NP_054720.1 | 1 - 2 | 116 | 64 |
NM_138619.3 | NP_619525.1 | 1 - 2 | 116 | 64 | |
XM_011524563.2 | XP_011522865.1 | 1 - 2 | 90 | 64 | |
XM_017024385.1 | XP_016879874.1 | 1 - 2 | 90 | 64 | |
XM_017024386.1 | XP_016879875.1 | 1 - 2 | 78 | 64 | |
XM_017024391.1 | XP_016879880.1 | 1 - 2 | 73 | 64 | |
GenBank mRNA | AF190864.1 | - | 1 - 2 | 50 | 64 |
AF219138.1 | - | 1 - 2 | 51 | 64 | |
AF219139.1 | - | 1 - 2 | 51 | 64 | |
AK291651.1 | - | 1 - 2 | 81 | 64 | |
AK291685.1 | - | 1 - 2 | 47 | 64 | |
AK293291.1 | - | 1 - 2 | 81 | 64 | |
AK302423.1 | - | 1 - 2 | 45 | 64 | |
AK304692.1 | - | 1 - 2 | 45 | 64 | |
BC063285.1 | - | 1 - 2 | 78 | 64 | |
BC070044.1 | - | 1 - 2 | 55 | 64 | |
D63876.1 | - | 1 - 2 | 50 | 64 | |
DA312660.1 | - | 1 - 2 | 116 | 64 |