Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92486899-92487415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
UniGene
Hs.164682
Gene Chromosome Location
Chr.7: 92487023-92528531 on Build GRCh38
Transcripts
NM_000466.2 NM_001282677.1 NM_001282678.1 XM_017012319.1

SNP Detail

SNP ID Chromosome Location
rs769772779 Chr.7: 92487029-92487029 on Build GRCh38
rs915801714 Chr.7: 92487140-92487140 on Build GRCh38
rs559962859 Chr.7: 92487034-92487034 on Build GRCh38
rs761817429 Chr.7: 92487192-92487192 on Build GRCh38
rs542790093 Chr.7: 92486955-92486955 on Build GRCh38
rs1056135929 Chr.7: 92487098-92487098 on Build GRCh38
rs886062500 Chr.7: 92487324-92487324 on Build GRCh38
rs1038387045 Chr.7: 92487241-92487241 on Build GRCh38
rs3180493 Chr.7: 92487024-92487024 on Build GRCh38
rs886062499 Chr.7: 92487216-92487216 on Build GRCh38
rs898507418 Chr.7: 92487249-92487249 on Build GRCh38
rs536884508 Chr.7: 92487130-92487130 on Build GRCh38
rs955676822 Chr.7: 92487319-92487319 on Build GRCh38
rs79220484 Chr.7: 92486954-92486954 on Build GRCh38
rs928392303 Chr.7: 92487217-92487217 on Build GRCh38
rs1045593763 Chr.7: 92487166-92487166 on Build GRCh38
rs528728841 Chr.7: 92487036-92487036 on Build GRCh38
rs552068263 Chr.7: 92487039-92487039 on Build GRCh38
rs893649883 Chr.7: 92487318-92487318 on Build GRCh38
rs972962611 Chr.7: 92487090-92487090 on Build GRCh38
rs1042346369 Chr.7: 92487374-92487374 on Build GRCh38
rs943129156 Chr.7: 92487258-92487258 on Build GRCh38
rs1003521008 Chr.7: 92486953-92486953 on Build GRCh38
rs10238260 Chr.7: 92486926-92486926 on Build GRCh38
rs548449757 Chr.7: 92487283-92487283 on Build GRCh38
rs955745000 Chr.7: 92486948-92486948 on Build GRCh38
rs1008875879 Chr.7: 92487323-92487323 on Build GRCh38
rs961491317 Chr.7: 92487068-92487068 on Build GRCh38
rs867076647 Chr.7: 92487124-92487124 on Build GRCh38
rs1029003656 Chr.7: 92487281-92487281 on Build GRCh38
rs1009362045 Chr.7: 92487373-92487373 on Build GRCh38
rs996832628 Chr.7: 92487277-92487277 on Build GRCh38
rs986956449 Chr.7: 92486952-92486952 on Build GRCh38
rs1004289500 Chr.7: 92487223-92487223 on Build GRCh38
rs376251385 Chr.7: 92487026-92487026 on Build GRCh38
rs1054059833 Chr.7: 92487278-92487278 on Build GRCh38
rs1021161458 Chr.7: 92487350-92487350 on Build GRCh38
rs967142323 Chr.7: 92487013-92487013 on Build GRCh38
rs886942535 Chr.7: 92487214-92487214 on Build GRCh38
rs748448769 Chr.7: 92487316-92487316 on Build GRCh38
rs35259480 Chr.7: 92487120-92487121 on Build GRCh38
rs763903553 Chr.7: 92486934-92486934 on Build GRCh38
rs138531138 Chr.7: 92487165-92487165 on Build GRCh38
rs910318700 Chr.7: 92487257-92487257 on Build GRCh38

View Full Product Details