The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015044.4 | NP_055859.1 | 5 - 6 | 554 | 85 |
XM_017023074.1 | XP_016878563.1 | 6 - 7 | 4243 | 85 | |
XM_017023075.1 | XP_016878564.1 | 5 - 6 | 557 | 85 | |
GenBank mRNA | AB029003.1 | - | 2 - 3 | 184 | 85 |
AF165531.1 | - | 5 - 6 | 492 | 85 | |
AF190863.1 | - | 5 - 6 | 498 | 85 | |
AF233522.1 | - | 5 - 6 | 497 | 85 | |
AF323754.1 | - | 5 - 6 | 496 | 85 | |
AK308425.1 | - | 5 - 6 | 541 | 85 | |
BC000284.1 | - | 5 - 6 | 536 | 85 |