Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKHD1
Entrez Gene ID
5314
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Aliases
ARPKD, FCYT, TIGM1
Chromosome Location
Chr.6: 51613291 - 52087625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138694.3 NP_619639.3 35 - 36 6031 94
NM_170724.2 NP_733842.2 35 - 36 6031 94
XM_011514680.2 XP_011512982.1 35 - 36 6375 94
XM_011514682.2 XP_011512984.1 35 - 36 5997 94
XM_011514683.2 XP_011512985.1 36 - 37 5355 94
XM_011514684.2 XP_011512986.1 27 - 28 5141 94
XM_011514685.1 XP_011512987.1 35 - 36 6031 94
XM_011514686.2 XP_011512988.1 35 - 36 6031 94
XM_011514687.1 XP_011512989.1 35 - 36 6031 94
XM_011514688.2 XP_011512990.1 35 - 36 5997 94
XM_017010944.1 XP_016866433.1 35 - 36 5857 94
XM_017010945.1 XP_016866434.1 34 - 35 5922 94
XM_017010946.1 XP_016866435.1 35 - 36 5997 94
XM_017010947.1 XP_016866436.1 33 - 34 5733 94
XM_017010948.1 XP_016866437.1 26 - 27 5815 94
XM_017010949.1 XP_016866438.1 17 - 18 4994 94
XM_017010950.1 XP_016866439.1 35 - 36 6031 94
XM_017010951.1 XP_016866440.1 35 - 36 5997 94
XM_017010952.1 XP_016866441.1 35 - 36 5997 94
GenBank mRNA AF480064.1 - 35 - 36 5977 94
AY074797.1 - 35 - 36 6031 94
AY092083.1 - 35 - 36 6017 94

View Full Product Details