The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AF490769.1 | - | 2 - 3 | 380 | 97 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | MAP3K7 C-terminal like | MAP3K7CL | C21orf7, HC21ORF7, TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4 | -18.2kb | Sense | Chr. - 29077471 - 29175889 on Build GRCh38 |
Downstream | BTB domain and CNC homolog 1 | BACH1 | BACH-1, BTBD24 | 69.8kb | Sense | Chr. - 29298795 - 29361896 on Build GRCh38 |