Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96171386-96171885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
UniGene
Hs.489190
Gene Chromosome Location
Chr.7: 96120220-96322147 on Build GRCh38
Transcripts
NM_001160210.1 NM_014251.2 NR_027662.1 XM_006715831.3 XM_011515727.2 XM_017011663.1 XM_017011664.1 XM_017011665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2865699 c.1230A>C p.T410T Chr.7: 96171472-96171472 on Build GRCh38
3883582 c.1221A>G p.I407M Chr.7: 96171481-96171481 on Build GRCh38
5615911 c.1219A>G p.I407V Chr.7: 96171483-96171483 on Build GRCh38
1453053 c.1194A>G p.L398L Chr.7: 96171508-96171508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571127452 Chr.7: 96171471-96171471 on Build GRCh38
rs569750564 Chr.7: 96171658-96171658 on Build GRCh38
rs954846610 Chr.7: 96171642-96171642 on Build GRCh38
rs528999533 Chr.7: 96171731-96171731 on Build GRCh38
rs767596108 Chr.7: 96171644-96171644 on Build GRCh38
rs751629925 Chr.7: 96171442-96171442 on Build GRCh38
rs926003284 Chr.7: 96171691-96171691 on Build GRCh38
rs970451407 Chr.7: 96171818-96171818 on Build GRCh38
rs371936313 Chr.7: 96171424-96171424 on Build GRCh38
rs776409965 Chr.7: 96171549-96171549 on Build GRCh38
rs555465094 Chr.7: 96171701-96171701 on Build GRCh38
rs759909171 Chr.7: 96171574-96171574 on Build GRCh38
rs755207755 Chr.7: 96171443-96171443 on Build GRCh38
rs201432251 Chr.7: 96171457-96171457 on Build GRCh38
rs990573739 Chr.7: 96171785-96171785 on Build GRCh38
rs149840781 Chr.7: 96171697-96171697 on Build GRCh38
rs2301629 Chr.7: 96171508-96171508 on Build GRCh38
rs756485341 Chr.7: 96171469-96171469 on Build GRCh38
rs777133632 Chr.7: 96171422-96171425 on Build GRCh38
rs755015021 Chr.7: 96171576-96171576 on Build GRCh38
rs369195484 Chr.7: 96171483-96171483 on Build GRCh38
rs144847839 Chr.7: 96171775-96171775 on Build GRCh38
rs907786458 Chr.7: 96171743-96171743 on Build GRCh38
rs572114737 Chr.7: 96171703-96171703 on Build GRCh38
rs890204198 Chr.7: 96171834-96171834 on Build GRCh38
rs762909695 Chr.7: 96171794-96171794 on Build GRCh38
rs949159861 Chr.7: 96171732-96171732 on Build GRCh38
rs377325226 Chr.7: 96171537-96171537 on Build GRCh38
rs547124508 Chr.7: 96171821-96171821 on Build GRCh38
rs760480472 Chr.7: 96171423-96171423 on Build GRCh38
rs1029178080 Chr.7: 96171605-96171605 on Build GRCh38
rs1010165385 Chr.7: 96171665-96171665 on Build GRCh38
rs765139878 Chr.7: 96171568-96171568 on Build GRCh38
rs150021522 Chr.7: 96171472-96171472 on Build GRCh38
rs1004549438 Chr.7: 96171506-96171506 on Build GRCh38
rs549957039 Chr.7: 96171428-96171428 on Build GRCh38
rs373876605 Chr.7: 96171505-96171505 on Build GRCh38
rs901200620 Chr.7: 96171661-96171661 on Build GRCh38
rs376581486 Chr.7: 96171427-96171427 on Build GRCh38
rs931225389 Chr.7: 96171597-96171597 on Build GRCh38
rs1046302376 Chr.7: 96171741-96171741 on Build GRCh38
rs996256932 Chr.7: 96171791-96171791 on Build GRCh38
rs556129855 Chr.7: 96171554-96171554 on Build GRCh38
rs979160155 Chr.7: 96171660-96171660 on Build GRCh38
rs759235789 Chr.7: 96171702-96171702 on Build GRCh38
rs775413889 Chr.7: 96171753-96171753 on Build GRCh38
rs779536772 Chr.7: 96171458-96171458 on Build GRCh38
rs59694807 Chr.7: 96171761-96171761 on Build GRCh38
rs201352939 Chr.7: 96171461-96171461 on Build GRCh38
rs995963314 Chr.7: 96171548-96171548 on Build GRCh38
rs889618864 Chr.7: 96171625-96171625 on Build GRCh38
rs774051691 Chr.7: 96171496-96171496 on Build GRCh38
rs760401530 Chr.7: 96171516-96171516 on Build GRCh38
rs753718735 Chr.7: 96171426-96171426 on Build GRCh38
rs1020453339 Chr.7: 96171650-96171650 on Build GRCh38
rs751616319 Chr.7: 96171572-96171572 on Build GRCh38

View Full Product Details