Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBX3
Entrez Gene ID
11335
Gene Name
chromobox 3
Gene Aliases
HECH, HP1-GAMMA, HP1Hs-gamma
Chromosome Location
Chr.7: 26201211 - 26213607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007276.4 NP_009207.2 1 - 2 355 130
GenBank mRNA BC000954.1 - 1 - 2 87 130
CV575477.1 - 1 - 2 381 130

View Full Product Details