The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022075.4 | NP_071358.1 | 8 - 9 | 1130 | 66 |
NM_181746.3 | NP_859530.1 | 8 - 9 | 1170 | 66 | |
XM_011509451.2 | XP_011507753.1 | 8 - 9 | 833 | 66 | |
XM_011509452.2 | XP_011507754.1 | 8 - 9 | 881 | 66 | |
GenBank mRNA | AF177338.2 | - | 8 - 9 | 859 | 66 |
AF189062.4 | - | 3 - 4 | 343 | 66 | |
AK001105.1 | - | 7 - 8 | 799 | 66 | |
AK290989.1 | - | 8 - 9 | 806 | 66 | |
AY091458.1 | - | 8 - 9 | 952 | 66 | |
BC001357.2 | - | 8 - 9 | 825 | 66 | |
BC010032.2 | - | 8 - 9 | 789 | 66 |