The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016038.2 | NP_057122.2 | 3 - 4 | 640 | 116 |
GenBank mRNA | AF151855.1 | - | 3 - 4 | 626 | 116 |
AK001779.1 | - | 3 - 4 | 640 | 116 | |
AK289609.1 | - | 3 - 4 | 645 | 116 | |
AY169963.1 | - | 3 - 4 | 640 | 116 | |
BC065700.1 | - | 3 - 4 | 610 | 116 | |
KU178629.1 | - | 3 - 4 | 456 | 116 | |
KU178630.1 | - | 2 - 3 | 326 | 116 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_001588.2 | 3 - 4 | 727 | 116 | |
NR_024109.1 | 2 - 3 | 605 | 116 | ||
NR_024110.1 | 3 - 4 | 727 | 116 | ||
NR_024111.1 | 2 - 3 | 605 | 116 | ||
GenBank mRNA | BQ441749.1 | - | 2 - 3 | 464 | 116 |
CB216674.1 | - | 1 - 2 | 156 | 116 | |
CN267170.1 | - | 2 - 3 | 490 | 116 | |
CX867149.1 | - | 3 - 4 | 619 | 116 |