Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
Chromosome Location
Chr.3: 3148490 - 3179717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173482.1 NP_001166953.1 9 - 10 1041 147
NM_016302.3 NP_057386.2 9 - 10 1044 147
XM_005265202.3 XP_005265259.1 8 - 9 925 147
XM_011533793.2 XP_011532095.1 10 - 11 1172 147
XM_011533794.2 XP_011532096.1 9 - 10 1043 147
GenBank mRNA AF117230.1 - 9 - 10 1008 146
AK027507.1 - 9 - 10 1022 147
AK058191.1 - 9 - 10 1018 147
AK123077.1 - 1 - 2 1417 147
AK301423.1 - 8 - 9 916 147
BC017419.1 - 9 - 10 1032 147
BC067811.1 - 10 - 11 1025 147
BX647351.1 - 1 - 2 3356 147
CR627060.1 - 5 - 6 3437 147

View Full Product Details