The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018352.2 | NP_060822.2 | 4 - 5 | 523 | 148 |
GenBank mRNA | AK002046.1 | - | 4 - 5 | 354 | 148 |
AK298235.1 | - | 4 - 5 | 409 | 148 | |
BC011842.2 | - | 4 - 5 | 340 | 148 | |
BU159489.1 | - | 4 - 5 | 523 | 148 |