Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDP1
Entrez Gene ID
55814
Gene Name
B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB
Gene Aliases
HSA238520, TAF3B1, TFC5, TFIIIB'', TFIIIB150, TFIIIB90, TFNR
Chromosome Location
Chr.5: 71455615 - 71578288 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018429.2 NP_060899.2 1 - 2 475 112
XM_011543511.2 XP_011541813.1 1 - 2 453 112
XM_011543512.2 XP_011541814.1 1 - 2 454 112
XM_017009630.1 XP_016865119.1 1 - 2 454 112
XM_017009631.1 XP_016865120.1 1 - 2 454 112
XM_017009632.1 XP_016865121.1 1 - 2 453 112
XM_017009633.1 XP_016865122.1 1 - 2 453 112
XM_017009634.1 XP_016865123.1 1 - 2 453 112
GenBank mRNA AB051476.3 - 1 - 2 222 112
AF176695.1 - 1 - 2 431 112
AF298151.1 - 1 - 2 301 112
AF298152.1 - 1 - 2 301 112
AJ238520.2 - 1 - 2 416 112
AK057871.1 - 2 - 3 376 112
BC032146.2 - 1 - 2 417 112
BC146792.1 - 1 - 2 222 112
BC161615.1 - 1 - 2 204 112
BP246483.1 - 1 - 2 475 112
BP347992.1 - 1 - 2 450 112

View Full Product Details