The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020824.3 | NP_065875.3 | 8 - 9 | 1015 | 88 |
XM_005252542.3 | XP_005252599.1 | 8 - 9 | 1083 | 88 | |
XM_011519602.2 | XP_011517904.1 | 8 - 9 | 2768 | 88 | |
XM_011519603.2 | XP_011517905.1 | 8 - 9 | 1175 | 88 | |
XM_011519604.2 | XP_011517906.1 | 8 - 9 | 991 | 88 | |
XM_011519606.2 | XP_011517908.1 | 5 - 6 | 2633 | 88 | |
XM_011519607.2 | XP_011517909.1 | 8 - 9 | 2768 | 88 | |
XM_017016459.1 | XP_016871948.1 | 8 - 9 | 1175 | 88 | |
GenBank mRNA | AB037845.2 | - | 7 - 8 | 488 | 88 |
AF480466.1 | - | 8 - 9 | 962 | 88 | |
AK307604.1 | - | 8 - 9 | 992 | 88 |