The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022460.3 | NP_071905.3 | 4 - 5 | 662 | 96 |
XM_017004696.1 | XP_016860185.1 | 4 - 5 | 665 | 96 | |
XM_017004697.1 | XP_016860186.1 | 4 - 5 | 665 | 96 | |
XM_017004698.1 | XP_016860187.1 | 4 - 5 | 665 | 96 | |
XM_017004699.1 | XP_016860188.1 | 4 - 5 | 717 | 96 | |
XM_017004700.1 | XP_016860189.1 | 4 - 5 | 761 | 96 | |
XM_017004701.1 | XP_016860190.1 | 4 - 5 | 665 | 96 | |
GenBank mRNA | AK314383.1 | - | 4 - 5 | 658 | 96 |
BC050636.1 | - | 4 - 5 | 642 | 96 |