The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030957.3 | NP_112219.3 | 3 - 4 | 395 | 81 |
XM_006722917.3 | XP_006722980.1 | 2 - 3 | 236 | 81 | |
XM_017027338.1 | XP_016882827.1 | 2 - 3 | 251 | 81 | |
GenBank mRNA | AB209515.1 | - | 2 - 3 | 226 | 81 |
BI822988.1 | - | 3 - 4 | 335 | 81 |