The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052950.3 | NP_443182.1 | 3 - 4 | 612 | 113 |
XM_011534914.1 | XP_011533216.1 | 3 - 4 | 503 | 113 | |
GenBank mRNA | AK309815.1 | - | 3 - 4 | 465 | 113 |
BC014004.2 | - | 3 - 4 | 452 | 113 |