Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIB2
- Entrez Gene ID
-
142678
- Gene Name
- mindbomb E3 ubiquitin protein ligase 2
- Gene Aliases
-
ZZANK1, ZZZ5
- Chromosome Location
-
Chr.1: 1615188 - 1630610 on Build GRCh38
View Full Product Details