The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207015.1 | NP_001193944.1 | 11 - 12 | 1529 | 77 |
NM_001207016.1 | NP_001193945.1 | 9 - 10 | 1319 | 77 | |
NM_002849.3 | NP_002840.2 | 12 - 13 | 2185 | 77 | |
NM_130846.2 | NP_570897.2 | 8 - 9 | 1451 | 77 | |
NR_073474.1 | 8 - 9 | 1521 | 77 | ||
XM_011538615.2 | XP_011536917.1 | 12 - 13 | 1781 | 77 | |
GenBank mRNA | AK091647.1 | - | 3 - 4 | 424 | 77 |
AK295951.1 | - | 11 - 12 | 1519 | 77 | |
AK304457.1 | - | 9 - 10 | 1278 | 77 | |
AK312376.1 | - | 12 - 13 | 2098 | 77 | |
BC110900.1 | - | 8 - 9 | 1148 | 77 | |
BC119777.1 | - | 9 - 10 | 1127 | 77 | |
BC122866.1 | - | 9 - 10 | 1127 | 77 | |
BX571751.1 | - | 8 - 9 | 1504 | 77 | |
CR749836.1 | - | 3 - 4 | 584 | 77 | |
D64053.1 | - | 12 - 13 | 2187 | 77 | |
U42361.1 | - | 12 - 13 | 1945 | 77 | |
U77916.1 | - | 9 - 10 | 1169 | 77 | |
U77917.1 | - | 8 - 9 | 1428 | 77 | |
X82635.1 | - | 8 - 9 | 1409 | 77 | |
Z79693.1 | - | 12 - 13 | 1856 | 77 |