Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHBDL3
Entrez Gene ID
162494
Gene Name
rhomboid like 3
Gene Aliases
RHBDL4, VRHO
Chromosome Location
Chr.17: 32266167 - 32324661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138328.2 NP_612201.1 7 - 8 896 69
XM_006721733.3 XP_006721796.1 6 - 7 873 69
XM_006721734.3 XP_006721797.1 6 - 7 1265 69
XM_011524443.2 XP_011522745.1 5 - 6 714 69
XM_017024279.1 XP_016879768.1 5 - 6 1539 69
GenBank mRNA AJ313480.1 - 7 - 8 896 69
AK124570.1 - 5 - 6 710 69
AK127490.1 - 1 - 2 55 69
BC100975.1 - 7 - 8 895 69
BC100977.1 - 6 - 7 872 69
BC100978.1 - 7 - 8 875 69

View Full Product Details