Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 18 - 19 4047 71
NM_138732.2 NP_620060.1 16 - 17 3927 71
NM_138734.2 NP_620063.1 2 - 3 959 71
XM_005274400.3 XP_005274457.1 17 - 18 3579 71
XM_005274401.3 XP_005274458.1 17 - 18 3579 71
XM_005274402.3 XP_005274459.1 16 - 17 3555 71
XM_011545370.1 XP_011543672.1 17 - 18 3609 71
XM_011545371.1 XP_011543673.1 17 - 18 3606 71
XM_011545373.1 XP_011543675.1 17 - 18 3588 71
XM_011545375.1 XP_011543677.1 16 - 17 3564 71
XM_011545385.1 XP_011543687.1 10 - 11 1954 71
XM_017018563.1 XP_016874052.1 16 - 17 3579 71
XM_017018564.1 XP_016874053.1 16 - 17 3576 71
XM_017018565.1 XP_016874054.1 16 - 17 3573 71
XM_017018566.1 XP_016874055.1 16 - 17 3537 71
XM_017018567.1 XP_016874056.1 17 - 18 3609 71
XM_017018568.1 XP_016874057.1 17 - 18 3600 71
XM_017018569.1 XP_016874058.1 15 - 16 3507 71
XM_017018570.1 XP_016874059.1 17 - 18 3609 71
XM_017018571.1 XP_016874060.1 17 - 18 3600 71
XM_017018572.1 XP_016874061.1 17 - 18 2904 71
XM_017018573.1 XP_016874062.1 16 - 17 3573 71
XM_017018574.1 XP_016874063.1 16 - 17 2874 71
XM_017018575.1 XP_016874064.1 16 - 17 2871 71
XM_017018576.1 XP_016874065.1 12 - 13 2519 71
XM_017018577.1 XP_016874066.1 10 - 11 1945 71
XM_017018578.1 XP_016874067.1 17 - 18 3600 71
GenBank mRNA AB023138.2 - 17 - 18 3904 71
AB035266.1 - 17 - 18 3699 71
AB177862.1 - 14 - 15 2751 71
BC150275.1 - 17 - 18 3904 71

View Full Product Details