Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location
Chr.9: 76611376 - 76906220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308047.1 NP_001294976.1 5 - 6 780 87
NM_001308048.1 NP_001294977.1 5 - 6 780 87
NM_015225.2 NP_056040.2 5 - 6 780 87
NR_131751.1 4 - 5 577 87
XM_005251745.1 XP_005251802.1 5 - 6 913 87
XM_005251746.1 XP_005251803.1 5 - 6 913 87
XM_005251748.1 XP_005251805.1 5 - 6 913 87
XM_005251750.1 XP_005251807.1 5 - 6 913 87
XM_005251751.1 XP_005251808.1 5 - 6 913 87
XM_005251754.1 XP_005251811.1 5 - 6 913 87
XM_006716982.1 XP_006717045.1 5 - 6 913 87
XM_006716983.1 XP_006717046.1 5 - 6 913 87
XM_006716984.2 XP_006717047.1 5 - 6 913 87
XM_006716985.1 XP_006717048.1 5 - 6 913 87
XM_006716986.1 XP_006717049.1 5 - 6 913 87
XM_011518323.1 XP_011516625.1 5 - 6 913 87
XM_011518326.2 XP_011516628.1 5 - 6 913 87
XM_011518327.1 XP_011516629.1 5 - 6 913 87
XM_011518328.2 XP_011516630.1 5 - 6 913 87
XM_017014345.1 XP_016869834.1 5 - 6 913 87
XM_017014346.1 XP_016869835.1 5 - 6 913 87
XM_017014347.1 XP_016869836.1 5 - 6 913 87
XM_017014348.1 XP_016869837.1 5 - 6 913 87
XM_017014349.1 XP_016869838.1 5 - 6 913 87
XM_017014350.1 XP_016869839.1 5 - 6 913 87
XM_017014351.1 XP_016869840.1 5 - 6 913 87
XM_017014352.1 XP_016869841.1 5 - 6 913 87
XM_017014353.1 XP_016869842.1 5 - 6 913 87
GenBank mRNA AB535152.1 - 5 - 6 656 87
BC019095.2 - 5 - 6 780 87
DA410068.1 - 3 - 4 396 87
FJ808772.1 - 5 - 6 734 87
FJ808773.1 - 4 - 5 531 87

View Full Product Details