The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006951.4 | NP_008882.2 | 1 - 2 | 595 | 82 |
NM_139052.2 | NP_620640.1 | 1 - 2 | 595 | 82 | |
GenBank mRNA | AK223615.1 | - | 1 - 2 | 567 | 82 |
AK291229.1 | - | 1 - 2 | 595 | 82 | |
BC052268.1 | - | 1 - 2 | 573 | 82 | |
BC136340.1 | - | 1 - 2 | 576 | 82 | |
BC136348.1 | - | 1 - 2 | 576 | 82 | |
U75309.1 | - | 1 - 2 | 561 | 82 | |
U80191.1 | - | 1 - 2 | 572 | 82 | |
X95525.1 | - | 1 - 2 | 581 | 82 |