Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C16orf71
Entrez Gene ID
146562
Gene Name
chromosome 16 open reading frame 71
Gene Aliases
-
Chromosome Location
Chr.16: 4734288 - 4749396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139170.2 NP_631909.2 8 - 9 1740 104
XM_005255144.3 XP_005255201.1 7 - 8 1469 104
XM_017022975.1 XP_016878464.1 8 - 9 1596 104
XM_017022976.1 XP_016878465.1 8 - 9 1358 104
GenBank mRNA AF447587.1 - 8 - 9 1714 104
AK125251.1 - 5 - 6 769 104
AK301794.1 - 8 - 9 1566 104
BC035024.1 - 8 - 9 1690 104

View Full Product Details