The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022917.4 | NP_075068.2 | 12 - 13 | 1708 | 73 |
NM_139235.3 | NP_631981.2 | 12 - 13 | 1708 | 73 | |
XM_017015044.1 | XP_016870533.1 | 12 - 13 | 1664 | 73 | |
GenBank mRNA | AF361079.1 | - | 12 - 13 | 1666 | 73 |
AF361080.1 | - | 12 - 13 | 1666 | 73 | |
AF361081.1 | - | 12 - 13 | 1666 | 73 | |
AK056559.1 | - | 4 - 5 | 422 | 73 | |
AK293969.1 | - | 11 - 12 | 1511 | 73 | |
BC008852.2 | - | 1 - 2 | 101 | 73 | |
BC030139.2 | - | 12 - 13 | 1612 | 73 |