Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCKBR
Entrez Gene ID
887
Gene Name
cholecystokinin B receptor
Gene Aliases
CCK-B, CCK2R, GASR
Chromosome Location
Chr.11: 6259611 - 6272127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176875.3 NP_795344.1 1 - 2 408 131
XM_005253210.1 XP_005253267.1 1 - 2 471 131
GenBank mRNA AF239668.1 - 1 - 2 156 131
AF441129.1 - 1 - 2 153 131
AK226020.1 - 1 - 2 221 131
AK292064.1 - 1 - 2 226 131
AK314266.1 - 1 - 2 186 131
AY029770.1 - 1 - 2 153 131
AY322551.1 - 1 - 2 153 131
BC000740.2 - 1 - 2 209 131
BT006789.1 - 1 - 2 153 131
CX762706.1 - 1 - 2 207 131
D13305.1 - 1 - 2 345 131
DA297178.1 - 1 - 2 471 131
L04473.1 - 1 - 2 153 131
L07746.1 - 1 - 2 315 131
L08112.1 - 1 - 2 153 131
S70057.1 - 1 - 2 338 131

View Full Product Details