The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003525.2 | NP_003516.1 | 1 - 1 | 351 | 116 |
GenBank mRNA | BC101653.1 | - | 1 - 1 | 411 | 116 |
BC101655.1 | - | 1 - 1 | 411 | 116 |