The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281432.1 | NP_001268361.1 | 2 - 3 | 197 | 110 |
NM_001281433.1 | NP_001268362.1 | 2 - 3 | 197 | 110 | |
NM_004773.3 | NP_004764.1 | 2 - 3 | 197 | 110 | |
GenBank mRNA | AF400652.1 | - | 2 - 3 | 189 | 110 |
AK290858.1 | - | 2 - 3 | 148 | 110 | |
AK310022.1 | - | 2 - 3 | 132 | 110 | |
AV763423.1 | - | - | 164 | 110 | |
BC017931.1 | - | 2 - 3 | 162 | 110 | |
BG032116.1 | - | 2 - 3 | 132 | 110 | |
BI767446.1 | - | 2 - 3 | 130 | 110 | |
HY045632.1 | - | 2 - 3 | 198 | 110 | |
L40410.1 | - | 2 - 3 | 115 | 110 |