The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306177.1 | NP_001293106.1 | 15 - 16 | 1831 | 119 |
NM_004840.2 | NP_004831.1 | 16 - 17 | 2293 | 119 | |
XM_011531412.2 | XP_011529714.1 | 17 - 18 | 2686 | 119 | |
XM_011531413.1 | XP_011529715.1 | 16 - 17 | 2617 | 119 | |
XM_011531415.2 | XP_011529717.1 | 16 - 17 | 2041 | 119 | |
XM_011531416.2 | XP_011529718.1 | 16 - 17 | 2304 | 119 | |
XM_011531417.1 | XP_011529719.1 | 15 - 16 | 2235 | 119 | |
XM_017029975.1 | XP_016885464.1 | 17 - 18 | 2698 | 119 | |
GenBank mRNA | AK291742.1 | - | 16 - 17 | 1836 | 119 |
AK294929.1 | - | 15 - 16 | 1637 | 119 | |
AK295709.1 | - | 16 - 17 | 1913 | 119 | |
BC033768.1 | - | 4 - 5 | 464 | 119 | |
BC039856.1 | - | 16 - 17 | 2293 | 119 | |
BC043505.1 | - | 15 - 16 | 1797 | 119 | |
BX537390.1 | - | 16 - 17 | 1898 | 119 | |
D13631.1 | - | 8 - 9 | 871 | 119 | |
D25304.1 | - | 16 - 17 | 1824 | 119 |