The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306177.1 | NP_001293106.1 | 19 - 20 | 2133 | 62 |
NM_004840.2 | NP_004831.1 | 20 - 21 | 2595 | 62 | |
XM_005262499.2 | XP_005262556.1 | 18 - 19 | 2760 | 62 | |
XM_011531412.2 | XP_011529714.1 | 21 - 22 | 2988 | 62 | |
XM_011531413.1 | XP_011529715.1 | 20 - 21 | 2919 | 62 | |
XM_011531414.1 | XP_011529716.1 | 19 - 20 | 2898 | 62 | |
XM_011531415.2 | XP_011529717.1 | 20 - 21 | 2343 | 62 | |
XM_011531416.2 | XP_011529718.1 | 20 - 21 | 2606 | 62 | |
XM_011531417.1 | XP_011529719.1 | 19 - 20 | 2537 | 62 | |
XM_017029975.1 | XP_016885464.1 | 21 - 22 | 3000 | 62 | |
XM_017029976.1 | XP_016885465.1 | 17 - 18 | 2378 | 62 | |
GenBank mRNA | AK291742.1 | - | 20 - 21 | 2138 | 62 |
AK294929.1 | - | 19 - 20 | 1939 | 62 | |
AK295709.1 | - | 20 - 21 | 2215 | 62 | |
BC033768.1 | - | 8 - 9 | 766 | 62 | |
BC039856.1 | - | 20 - 21 | 2595 | 62 | |
BC043505.1 | - | 19 - 20 | 2099 | 62 | |
BX537390.1 | - | 20 - 21 | 2200 | 62 | |
D13631.1 | - | 12 - 13 | 1173 | 62 | |
D25304.1 | - | 20 - 21 | 2126 | 62 |