Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1770775-1771290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3 XM_005266021.3 XM_005266022.1 XM_005266023.1 XM_011534745.1 XM_011534746.2 XM_011534747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122201 c.53A>G p.Y18C Chr.8: 1771107-1771107 on Build GRCh38
3898493 c.84G>T p.M28I Chr.8: 1771138-1771138 on Build GRCh38
110575 c.84G>A p.M28I Chr.8: 1771138-1771138 on Build GRCh38
5096283 c.78G>A p.T26T Chr.8: 1771132-1771132 on Build GRCh38
5636188 c.112G>A p.V38I Chr.8: 1771166-1771166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763583156 Chr.8: 1771120-1771120 on Build GRCh38
rs796069556 Chr.8: 1771228-1771228 on Build GRCh38
rs760919296 Chr.8: 1771185-1771185 on Build GRCh38
rs746626264 Chr.8: 1771208-1771208 on Build GRCh38
rs918232547 Chr.8: 1771076-1771076 on Build GRCh38
rs148763945 Chr.8: 1771138-1771138 on Build GRCh38
rs546337704 Chr.8: 1770857-1770857 on Build GRCh38
rs1057516867 Chr.8: 1771101-1771101 on Build GRCh38
rs374723418 Chr.8: 1770942-1770942 on Build GRCh38
rs372268977 Chr.8: 1771215-1771215 on Build GRCh38
rs757016989 Chr.8: 1771211-1771211 on Build GRCh38
rs770291875 Chr.8: 1771119-1771119 on Build GRCh38
rs886996594 Chr.8: 1771000-1771000 on Build GRCh38
rs758068226 Chr.8: 1771148-1771148 on Build GRCh38
rs555757531 Chr.8: 1771201-1771201 on Build GRCh38
rs746677418 Chr.8: 1771162-1771162 on Build GRCh38
rs753737213 Chr.8: 1771141-1771141 on Build GRCh38
rs762984374 Chr.8: 1771265-1771265 on Build GRCh38
rs796052361 Chr.8: 1770933-1770933 on Build GRCh38
rs767218890 Chr.8: 1771061-1771061 on Build GRCh38
rs958944574 Chr.8: 1771029-1771029 on Build GRCh38
rs541840243 Chr.8: 1771181-1771181 on Build GRCh38
rs924793834 Chr.8: 1771090-1771090 on Build GRCh38
rs536738656 Chr.8: 1771071-1771071 on Build GRCh38
rs137852883 Chr.8: 1771142-1771142 on Build GRCh38
rs768159414 Chr.8: 1771060-1771060 on Build GRCh38
rs773997032 Chr.8: 1771063-1771063 on Build GRCh38
rs1040664086 Chr.8: 1770915-1770915 on Build GRCh38
rs781017885 Chr.8: 1771077-1771077 on Build GRCh38
rs1003476629 Chr.8: 1771001-1771001 on Build GRCh38
rs754933058 Chr.8: 1771192-1771192 on Build GRCh38
rs1039856226 Chr.8: 1770898-1770898 on Build GRCh38
rs374522261 Chr.8: 1771009-1771009 on Build GRCh38
rs386834129 Chr.8: 1771100-1771100 on Build GRCh38
rs750647318 Chr.8: 1771127-1771127 on Build GRCh38
rs753430388 Chr.8: 1771165-1771165 on Build GRCh38
rs779525549 Chr.8: 1771050-1771050 on Build GRCh38
rs763671368 Chr.8: 1771267-1771267 on Build GRCh38
rs376382403 Chr.8: 1771144-1771144 on Build GRCh38
rs142104002 Chr.8: 1771107-1771107 on Build GRCh38
rs148754057 Chr.8: 1770896-1770896 on Build GRCh38
rs953594605 Chr.8: 1771046-1771046 on Build GRCh38
rs796052362 Chr.8: 1771054-1771054 on Build GRCh38
rs886043652 Chr.8: 1771101-1771101 on Build GRCh38
rs924823645 Chr.8: 1770810-1770810 on Build GRCh38
rs745784815 Chr.8: 1771032-1771032 on Build GRCh38
rs386834139 Chr.8: 1771142-1771142 on Build GRCh38
rs746305254 Chr.8: 1771117-1771117 on Build GRCh38
rs933753788 Chr.8: 1771132-1771132 on Build GRCh38
rs114189810 Chr.8: 1770928-1770928 on Build GRCh38
rs764099428 Chr.8: 1771073-1771073 on Build GRCh38
rs201327850 Chr.8: 1771241-1771241 on Build GRCh38
rs757793660 Chr.8: 1771203-1771203 on Build GRCh38
rs543839915 Chr.8: 1771014-1771014 on Build GRCh38
rs370199508 Chr.8: 1771166-1771166 on Build GRCh38
rs767698401 Chr.8: 1771126-1771126 on Build GRCh38
rs368365607 Chr.8: 1771261-1771261 on Build GRCh38
rs542253440 Chr.8: 1770893-1770893 on Build GRCh38
rs751762914 Chr.8: 1771075-1771075 on Build GRCh38
rs766132197 Chr.8: 1771189-1771189 on Build GRCh38
rs374597781 Chr.8: 1771069-1771069 on Build GRCh38
rs762079123 Chr.8: 1771125-1771125 on Build GRCh38
rs796052364 Chr.8: 1771230-1771230 on Build GRCh38
rs566084111 Chr.8: 1770984-1770984 on Build GRCh38
rs993863249 Chr.8: 1770938-1770938 on Build GRCh38
rs148668081 Chr.8: 1771104-1771104 on Build GRCh38
rs777453297 Chr.8: 1771205-1771205 on Build GRCh38
rs386834124 Chr.8: 1771263-1771263 on Build GRCh38
rs192196274 Chr.8: 1771180-1771180 on Build GRCh38
rs373957283 Chr.8: 1771254-1771254 on Build GRCh38
rs386834137 Chr.8: 1771120-1771120 on Build GRCh38
rs1053427402 Chr.8: 1770865-1770865 on Build GRCh38
rs1048152901 Chr.8: 1770947-1770947 on Build GRCh38
rs756041356 Chr.8: 1771094-1771094 on Build GRCh38
rs765158624 Chr.8: 1771194-1771194 on Build GRCh38
rs532828506 Chr.8: 1770955-1770955 on Build GRCh38
rs150754905 Chr.8: 1771153-1771153 on Build GRCh38
rs766183817 Chr.8: 1771139-1771139 on Build GRCh38
rs386834123 Chr.8: 1771234-1771236 on Build GRCh38
rs776409703 Chr.8: 1771260-1771260 on Build GRCh38
rs749651452 Chr.8: 1771113-1771113 on Build GRCh38
rs888272793 Chr.8: 1770964-1770964 on Build GRCh38
rs989562026 Chr.8: 1771025-1771025 on Build GRCh38
rs201670636 Chr.8: 1771171-1771171 on Build GRCh38
rs573279536 Chr.8: 1770879-1770879 on Build GRCh38
rs370711548 Chr.8: 1771015-1771015 on Build GRCh38
rs780876758 Chr.8: 1771028-1771028 on Build GRCh38
rs755846977 Chr.8: 1771131-1771131 on Build GRCh38
rs957551479 Chr.8: 1771095-1771095 on Build GRCh38
rs942360618 Chr.8: 1771110-1771110 on Build GRCh38
rs769069643 Chr.8: 1771214-1771214 on Build GRCh38
rs769770739 Chr.8: 1771044-1771044 on Build GRCh38
rs779899858 Chr.8: 1771099-1771099 on Build GRCh38
rs770240445 Chr.8: 1771163-1771163 on Build GRCh38
rs104894064 Chr.8: 1771124-1771124 on Build GRCh38
rs4875957 Chr.8: 1770805-1770805 on Build GRCh38
rs1030962349 Chr.8: 1771147-1771147 on Build GRCh38
rs147181589 Chr.8: 1771065-1771065 on Build GRCh38
rs34238807 Chr.8: 1771115-1771116 on Build GRCh38
rs863224859 Chr.8: 1771266-1771266 on Build GRCh38
rs989594643 Chr.8: 1770872-1770872 on Build GRCh38
rs758683794 Chr.8: 1771010-1771010 on Build GRCh38
rs771203701 Chr.8: 1771258-1771258 on Build GRCh38
rs765097897 Chr.8: 1771262-1771262 on Build GRCh38
rs557682976 Chr.8: 1771020-1771020 on Build GRCh38
rs750274455 Chr.8: 1771081-1771081 on Build GRCh38
rs200999640 Chr.8: 1771070-1771070 on Build GRCh38
rs191347799 Chr.8: 1770874-1770874 on Build GRCh38
rs564362499 Chr.8: 1770946-1770946 on Build GRCh38
rs780551031 Chr.8: 1771213-1771213 on Build GRCh38
rs914874692 Chr.8: 1770873-1770873 on Build GRCh38
rs184346819 Chr.8: 1770965-1770965 on Build GRCh38
rs575820157 Chr.8: 1771255-1771255 on Build GRCh38
rs143730802 Chr.8: 1771055-1771055 on Build GRCh38
rs147267153 Chr.8: 1770837-1770837 on Build GRCh38
rs923957402 Chr.8: 1770818-1770818 on Build GRCh38
rs866717923 Chr.8: 1771206-1771206 on Build GRCh38
rs368792081 Chr.8: 1771066-1771066 on Build GRCh38
rs777981149 Chr.8: 1771012-1771012 on Build GRCh38

View Full Product Details