Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1782446-1782959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3

SNP Detail

SNP ID Chromosome Location
rs548356940 Chr.8: 1782497-1782497 on Build GRCh38
rs576501229 Chr.8: 1782708-1782708 on Build GRCh38
rs910230056 Chr.8: 1782784-1782784 on Build GRCh38
rs538235635 Chr.8: 1782586-1782586 on Build GRCh38
rs1025061073 Chr.8: 1782707-1782707 on Build GRCh38
rs887669869 Chr.8: 1782844-1782844 on Build GRCh38
rs977555321 Chr.8: 1782666-1782666 on Build GRCh38
rs1041894827 Chr.8: 1782503-1782503 on Build GRCh38
rs115698196 Chr.8: 1782529-1782529 on Build GRCh38
rs1008723915 Chr.8: 1782550-1782550 on Build GRCh38
rs370222132 Chr.8: 1782488-1782488 on Build GRCh38
rs750883483 Chr.8: 1782753-1782753 on Build GRCh38
rs567865373 Chr.8: 1782505-1782505 on Build GRCh38
rs769022013 Chr.8: 1782618-1782618 on Build GRCh38
rs78155116 Chr.8: 1782528-1782528 on Build GRCh38
rs917317381 Chr.8: 1782654-1782654 on Build GRCh38
rs1040200609 Chr.8: 1782562-1782562 on Build GRCh38
rs565289128 Chr.8: 1782734-1782734 on Build GRCh38
rs1048143993 Chr.8: 1782605-1782605 on Build GRCh38
rs569623702 Chr.8: 1782884-1782884 on Build GRCh38
rs545037566 Chr.8: 1782731-1782731 on Build GRCh38
rs534613482 Chr.8: 1782657-1782657 on Build GRCh38
rs929160717 Chr.8: 1782817-1782817 on Build GRCh38
rs924297821 Chr.8: 1782498-1782498 on Build GRCh38
rs966889919 Chr.8: 1782720-1782720 on Build GRCh38
rs530303738 Chr.8: 1782853-1782853 on Build GRCh38
rs935698172 Chr.8: 1782506-1782506 on Build GRCh38
rs943064474 Chr.8: 1782790-1782790 on Build GRCh38
rs991494035 Chr.8: 1782491-1782491 on Build GRCh38
rs73672963 Chr.8: 1782702-1782702 on Build GRCh38
rs993387391 Chr.8: 1782601-1782601 on Build GRCh38
rs950159234 Chr.8: 1782658-1782658 on Build GRCh38
rs763497437 Chr.8: 1782760-1782760 on Build GRCh38
rs748731042 Chr.8: 1782575-1782575 on Build GRCh38
rs1013631851 Chr.8: 1782704-1782704 on Build GRCh38
rs73541034 Chr.8: 1782527-1782527 on Build GRCh38
rs185961111 Chr.8: 1782793-1782793 on Build GRCh38
rs761807438 Chr.8: 1782622-1782622 on Build GRCh38
rs1047624492 Chr.8: 1782838-1782838 on Build GRCh38
rs532249826 Chr.8: 1782909-1782909 on Build GRCh38
rs950189875 Chr.8: 1782499-1782499 on Build GRCh38
rs558545114 Chr.8: 1782598-1782598 on Build GRCh38
rs550380222 Chr.8: 1782881-1782881 on Build GRCh38
rs766695317 Chr.8: 1782786-1782786 on Build GRCh38
rs1012851010 Chr.8: 1782906-1782906 on Build GRCh38
rs1006151090 Chr.8: 1782872-1782872 on Build GRCh38
rs895154606 Chr.8: 1782907-1782907 on Build GRCh38
rs751948401 Chr.8: 1782808-1782808 on Build GRCh38
rs901683560 Chr.8: 1782571-1782571 on Build GRCh38
rs879526822 Chr.8: 1782701-1782701 on Build GRCh38
rs574281845 Chr.8: 1782692-1782692 on Build GRCh38
rs900102401 Chr.8: 1782722-1782722 on Build GRCh38
rs978359774 Chr.8: 1782725-1782725 on Build GRCh38
rs775467130 Chr.8: 1782559-1782559 on Build GRCh38
rs1014753298 Chr.8: 1782828-1782828 on Build GRCh38
rs558468255 Chr.8: 1782619-1782619 on Build GRCh38
rs1055079538 Chr.8: 1782718-1782718 on Build GRCh38
rs771972051 Chr.8: 1782539-1782539 on Build GRCh38
rs961985725 Chr.8: 1782594-1782594 on Build GRCh38
rs936288901 Chr.8: 1782521-1782521 on Build GRCh38
rs957041812 Chr.8: 1782746-1782746 on Build GRCh38
rs890222920 Chr.8: 1782543-1782543 on Build GRCh38
rs554488671 Chr.8: 1782677-1782677 on Build GRCh38
rs973692886 Chr.8: 1782596-1782596 on Build GRCh38
rs777303692 Chr.8: 1782908-1782908 on Build GRCh38
rs868159911 Chr.8: 1782938-1782938 on Build GRCh38
rs945600082 Chr.8: 1782482-1782482 on Build GRCh38
rs1033661694 Chr.8: 1782662-1782662 on Build GRCh38
rs561610018 Chr.8: 1782848-1782848 on Build GRCh38
rs771531967 Chr.8: 1782579-1782579 on Build GRCh38
rs113843633 Chr.8: 1782778-1782778 on Build GRCh38
rs544983784 Chr.8: 1782826-1782826 on Build GRCh38
rs1053450000 Chr.8: 1782522-1782522 on Build GRCh38

View Full Product Details