The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014182.4 | NP_054901.1 | 3 - 4 | 419 | 99 |
GenBank mRNA | AB064961.1 | - | 3 - 4 | 373 | 99 |
AF161509.1 | - | 3 - 4 | 380 | 99 | |
AF173375.1 | - | 3 - 4 | 389 | 99 | |
AF395705.1 | - | 2 - 3 | 334 | 99 | |
AF395706.1 | - | 2 - 3 | 339 | 99 | |
AF395707.1 | - | 3 - 4 | 516 | 99 | |
AK223180.1 | - | 3 - 4 | 360 | 99 | |
AK314046.1 | - | 3 - 4 | 400 | 99 | |
BC012543.2 | - | 2 - 3 | 329 | 99 |