Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMCX2
Entrez Gene ID
9823
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
ALEX2, GASP9
Chromosome Location
Chr.X: 101655281 - 101659891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014782.6 NP_055597.1 1 - 2 133 112
XM_017029994.1 XP_016885483.1 1 - 2 92 112
GenBank mRNA BC012541.1 - 1 - 2 89 112

View Full Product Details