Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF12
Entrez Gene ID
25853
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
CT102, KIAA1892, TCC52, WDR40A
Chromosome Location
Chr.9: 34086383 - 34126773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015397.3 NP_056212.1 3 - 4 888 79
XM_011517843.1 XP_011516145.1 3 - 4 715 79
GenBank mRNA AB067479.1 - 3 - 4 808 79
AK000354.1 - 3 - 4 666 79
AK292935.1 - 3 - 4 694 79
BC008893.2 - 2 - 3 451 79
BC063823.1 - 3 - 4 806 79
DQ518909.1 - 3 - 4 808 79

View Full Product Details